2-Brom-6-chlor-4-(trifluormethyl)anilin

Lieferant: FLUOROCHEM

Synonyme: 4-Amino-3-brom-5-chlor-α,α,α-trifluortoluol, 4-Amino-3-brom-5-chlorbenzotrifluorid

CFF536.5 CFF536.10 004163-25G 004163-5G
CHMPCFF536.5EA 427 EUR
CHMPCFF536.5 CHMPCFF536.10 FLUO004163-25G FLUO004163-5G
2-Brom-6-chlor-4-(trifluormethyl)anilin
2-Brom-6-chlor-4-(trifluormethyl)anilin
Formel: C₇H₄BrClF₃N
Molecular Weight: 274.47 g/mol
Siedepunkt: 71…72 °C (0.25 mmHg)
Schmelzpunkt: 29…32 °C
MDL: MFCD00042180
CAS-Nummer: 109919-26-8
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR