ω-Brom-3-cyanacetophenon

Lieferant: FLUOROCHEM

CFF155.2.5 CFF155.5 CFF155.10 018078-1G 018078-5G
CHMPCFF155.2.5EA 401 EUR
CHMPCFF155.2.5 CHMPCFF155.5 CHMPCFF155.10 FLUO018078-1G FLUO018078-5G
ω-Brom-3-cyanacetophenon
ω-Brom-3-cyanacetophenon
Formel: C₉H₆BrNO
MDL: MFCD00833267
CAS-Nummer: 50916-55-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR