ω-Brom-4-cyanacetophenon
Lieferant: Thermo Scientific
SPB02347.0001
SPB02347.0010
SPB02347.1
SPB02347.10
MAYBSPB02347.0001EA
0
EUR
MAYBSPB02347.0001
MAYBSPB02347.0010
MAYBSPB02347.1
MAYBSPB02347.10
ω-Brom-4-cyanacetophenon
ω-Brom-4-cyanacetophenon
Formel:
C₉H₆BrNO Siedepunkt: 314 °C (1013 hPa) Schmelzpunkt: 92…94 °C |
MDL:
MFCD00052931 CAS-Nummer: 20099-89-2 UN: 2923 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...