ω-Brom-4-cyanacetophenon
Lieferant: FLUOROCHEM
Synonyme:
2-Brom-4'-cyanacetophenon, 4-Cyanphenacylbromid, Brommethyl-p-cyanphenylketon, 4-(Bromacetyl)benzonitril, α-Brom-4-cyanacetophenon
CFF524.5
CFF524.10
018079-1G
018079-5G
CHMPCFF524.5EA
481
EUR
CHMPCFF524.5
CHMPCFF524.10
FLUO018079-1G
FLUO018079-5G
ω-Brom-4-cyanacetophenon
ω-Brom-4-cyanacetophenon
Formel:
C₉H₆BrNO Molecular Weight: 224,06 g/mol Siedepunkt: 314 °C (1013 hPa) Schmelzpunkt: 92…94 °C |
MDL:
MFCD00052931 CAS-Nummer: 20099-89-2 UN: 2923 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...