ω-Brom-3-nitroacetophenon 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2-Brom-3'-nitroacetophenon, Brommethyl-m-nitrophenylketon, 3-Nitrophenacylbromid, α-Brom-3-nitroacetophenon
137470250
137470050
ACRO137470250EA
65.2
EUR
ACRO137470250
ACRO137470050
ω-Brom-3-nitroacetophenon 97%
ω-Brom-3-nitroacetophenon
Formel:
C₈H₆BrNO₃ Schmelzpunkt: 90…94 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00024512 CAS-Nummer: 2227-64-7 EINECS: 218-764-0 UN: 3261 ADR: 8,II |
Spezifikation Testergebnisse
Appearance | Yellow-green to beige to light orange Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 90 to 94 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...