ω-Brom-4-nitroacetophenon, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Brom-4'-nitroacetophenon, 4-Nitrophenacylbromid, Brommethyl-p-nitrophenylketon, α-Brom-4-nitroacetophenon
M-Clarity™ Qualitätsstufe = MQ200
245615-10G
245615-50G
SIAL245615-10GEA
62.8
EUR
SIAL245615-10G
SIAL245615-50G
ω-Brom-4-nitroacetophenon, Sigma-Aldrich®
ω-Brom-4-nitroacetophenon
Formel:
C₈H₆BrNO₃ Molecular Weight: 244.04 g/mol Schmelzpunkt: 100 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007356 CAS-Nummer: 99-81-0 EINECS: 202-789-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...