2'-Brom-4'-fluoracetophenon

Lieferant: FLUOROCHEM

002686-5G 002686-25G
FLUO002686-5GEA 87.9 EUR
FLUO002686-5G FLUO002686-25G
2'-Brom-4'-fluoracetophenon
2'-Brom-4'-fluoracetophenon
Formel: C₈H₆BrFO
MDL: MFCD00077464
CAS-Nummer: 1006-39-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR