2,6-Dibrom-p-toluidin

Lieferant: Thermo Scientific

Synonyme: 2,6-Dibrom-4-methylanilin, 4-Amino-3,5-dibromtoluol

RH00021.1 RH00021.10 RH00021.50
MAYBRH00021.1EA 0 EUR
MAYBRH00021.1 MAYBRH00021.10 MAYBRH00021.50
2,6-Dibrom-p-toluidin
2,6-Dibrom-p-toluidin
Formel: C₇H₇Br₂N
Molecular Weight: 264,95 g/mol
Siedepunkt: 296 °C (1013 hPa)
Schmelzpunkt: 79 °C
MDL: MFCD00007641
CAS-Nummer: 6968-24-7
EINECS: 230-182-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR