4'-Chlor-3'-fluoracetophenon ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-(4-Chloro-3-fluorophenyl)ethanone
H26373.03
H26373.06
H26373.14
H26373.03EA
67
EUR
H26373.03
H26373.06
H26373.14
4'-Chlor-3'-fluoracetophenon ≥97%
4'-Chlor-3'-fluoracetophenon
Purity: 97%
CAS No.: 151945-84-5
Molecular Formula: C8H6ClFO
Formula Weight: 172.59
MDL No.: MFCD04115858
Formel:
C₈H₆ClFO Molecular Weight: 172,59 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115858 CAS-Nummer: 151945-84-5 EINECS: None |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...