4'-Chlor-3'-fluoracetophenon

Lieferant: CHEMPUR

FL13798.POR 013798-1G 013798-5G
CHMPFL13798.POREA 0 EUR
CHMPFL13798.POR FLUO013798-1G FLUO013798-5G
4'-Chlor-3'-fluoracetophenon
4'-Chlor-3'-fluoracetophenon
Formel: C₈H₆ClFO
MDL: MFCD04115858
CAS-Nummer: 151945-84-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR