2-Chlorbenzylisothiocyanat ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
1-Chloro-2-(isothiocyanatomethyl)benzene
L10546.03
L10546.06
L10546.03EA
23
EUR
L10546.03
L10546.06
2-Chlorbenzylisothiocyanat ≥97%
2-Chlorbenzylisothiocyanat
Purity: 97%
CAS No.: 18967-44-7
Molecular Formula: C8H6ClNS
Formula Weight: 183.66
Storage Temperature: 2°C to 8°C
Refractive Index: 1.6140
Odour: Irritating
Boiling Point: 137-138°/6mm
Beilstein Reference: 2936688
UN No.: 2922
MDL No.: MFCD00041110
Formel:
C₈H₆ClNS Siedepunkt: 137…138 °C (6 mmHg) Dichte: 1,149 |
MDL:
MFCD00041110 CAS-Nummer: 18967-44-7 ADR: 8,III |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...