2',3',4'-Trihydroxyacetophenon ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-(2,3,4-Trihydroxyphenyl)ethanone, Gallacetophenone
A14364.14
A14364.06
A14364.22
A14364.14EA
101
EUR
A14364.14
A14364.06
A14364.22
2',3',4'-Trihydroxyacetophenon ≥98%
2',3',4'-Trihydroxyacetophenon
Purity: 98%
CAS No.: 528-21-2
Molecular Formula: C8H8O4
Formula Weight: 168.15
Melting Point: 169-172°
Merck Reference: 14,4342
MDL No.: MFCD00002193
Formel:
C₈H₈O₄ Molecular Weight: 168,15 g/mol Schmelzpunkt: 169…172 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002193 CAS-Nummer: 528-21-2 EINECS: 208-430-2 Merck Index: 14,4342 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...