2'-Acetoxyacetophenon ≥98%

Lieferant: Thermo Scientific

Synonyme: o-Acetylphenylacetat, 2-Acetylphenylacetat, Essigsäure(2-acetylphenyl)ester

B21624.06 B21624.14
B21624.06EA 34.1 EUR
B21624.06 B21624.14
2'-Acetoxyacetophenon ≥98%
2'-Acetoxyacetophenon

Purity: 98% CAS No.: 7250-94-4 Molecular Formula: C10H10O3 Formula Weight: 178.19 Melting Point: 88-91° Boiling Point: 118°/19mm MDL No.: MFCD00017223

Formel: C₁₀H₁₀O₃
Molecular Weight: 178,19 g/mol
Siedepunkt: 118 °C (19 mmHg)
Schmelzpunkt: 88…91 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017223
CAS-Nummer: 7250-94-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR