4'-Acetoxyacetophenon ≥99%
Lieferant: Thermo Scientific
Synonyme:
4-Acetylphenylacetat, p-Acetylphenylacetat, Essigsäure(4-acetylphenyl)ester
B22284.14
B22284.22
B22284.14EA
24.1
EUR
B22284.14
B22284.22
4'-Acetoxyacetophenon ≥99%
4'-Acetoxyacetophenon
Purity: 99%
CAS No.: 13031-43-1
Molecular Formula: C10H10O3
Formula Weight: 178.19
Melting Point: 51-54°
Boiling Point: 165-170°/14mm
MDL No.: MFCD00017229
Formel:
C₁₀H₁₀O₃ Molecular Weight: 178,19 g/mol Siedepunkt: 165…170 °C (14 mmHg) Schmelzpunkt: 51…54 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017229 CAS-Nummer: 13031-43-1 EINECS: 235-894-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...