ω,ω-Difluoracetophenon 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2,2-Difluoracetophenon, α,α-Difluoracetophenon

H25750.03 H25750.MD H25750.06
H25750.03EA 117 EUR
H25750.03 H25750.MD H25750.06
ω,ω-Difluoracetophenon 95%
ω,ω-Difluoracetophenon

Purity: 95% CAS No.: 395-01-7 Molecular Formula: C8H6F2O Formula Weight: 156.13 Refractive Index: 1.4955 Odour: Characteristic Boiling Point: 53-54°/12mm MDL No.: MFCD06246879

Formel: C₈H₆F₂O
Molecular Weight: 156.13 g/mol
Siedepunkt: 53…54 °C (12 mmHg)
Schmelzpunkt: 48…50 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06246879
CAS-Nummer: 395-01-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR