3'-Fluor-4'-hydroxyacetophenon
Lieferant: BIONET RESEARCH
Synonyme:
1-(3-Fluoro-4-hydroxyphenyl)-1-ethanone
3X-0862.1
3X-0862.10
3X-0862.5
3X-0862.500
BIOT3X-0862.1EA
0
EUR
BIOT3X-0862.1
BIOT3X-0862.10
BIOT3X-0862.5
BIOT3X-0862.500
3'-Fluor-4'-hydroxyacetophenon
3'-Fluor-4'-hydroxyacetophenon
Formel:
C₈H₇FO₂ Molecular Weight: 154,14 g/mol |
MDL:
MFCD00215840 CAS-Nummer: 403-14-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...