5'-Fluor-2'-hydroxyacetophenon

Lieferant: FLUOROCHEM

Synonyme: 1-(5-Fluoro-2-hydroxyphenyl)ethanone

001235-1G 001235-5G 001235-25G
FLUO001235-1GEA 0 EUR
FLUO001235-1G FLUO001235-5G FLUO001235-25G
5'-Fluor-2'-hydroxyacetophenon
5'-Fluor-2'-hydroxyacetophenon
Formel: C₈H₇FO₂
Molecular Weight: 154.14 g/mol
Siedepunkt: 182
Schmelzpunkt: 56
MDL: MFCD00011668
CAS-Nummer: 394-32-1
EINECS: 206-893-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR