4'-Fluor-2'-hydroxyacetophenon
Lieferant: FLUOROCHEM
Synonyme:
4-Fluoro-2-hydroxyacetophenone
003064-5G
003064-25G
FLUO003064-5GEA
76.2
EUR
FLUO003064-5G
FLUO003064-25G
4'-Fluor-2'-hydroxyacetophenon
4'-Fluor-2'-hydroxyacetophenon
Formel:
C₈H₇FO₂ Molecular Weight: 154.14 g/mol |
MDL:
MFCD00143203 CAS-Nummer: 1481-27-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...