2-Fluor-3-(trifluormethylphenyl)isocyanat

Lieferant: Thermo Scientific

AC31933.250 AC31933.1
MAYBAC31933.250EA 0 EUR
MAYBAC31933.250 MAYBAC31933.1
2-Fluor-3-(trifluormethylphenyl)isocyanat
2-Fluor-3-(trifluormethylphenyl)isocyanat
Formel: C₈H₃F₄NO
Molecular Weight: 205,11 g/mol
CAS-Nummer: 190774-52-8
UN: 2922
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR