2-(2-Methoxyphenoxy)-5-(trifluormethyl)anilin
Lieferant: Thermo Scientific
BTB04252.1
BTB04252.10
MAYBBTB04252.1EA
0
EUR
MAYBBTB04252.1
MAYBBTB04252.10
2-(2-Methoxyphenoxy)-5-(trifluormethyl)anilin
2-(2-Methoxyphenoxy)-5-(trifluormethyl)anilin
Formel:
C₁₄H₁₂F₃NO₂ Molecular Weight: 283,25 g/mol |
MDL:
MFCD00052419 CAS-Nummer: 175135-08-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...