2-(2-Methoxyphenoxy)-5-(trifluormethyl)anilin

Lieferant: FLUOROCHEM

003999-1G 003999-5G
FLUO003999-1GEA 96.6 EUR
FLUO003999-1G FLUO003999-5G
2-(2-Methoxyphenoxy)-5-(trifluormethyl)anilin
2-(2-Methoxyphenoxy)-5-(trifluormethyl)anilin
Formel: C₁₄H₁₂F₃NO₂
Molecular Weight: 283.25 g/mol
MDL: MFCD00052419
CAS-Nummer: 175135-08-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR