4-Iodcumol ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Iodisopropylbenzol, 2-(4-Iodphenyl)propan
B25476.06
B25476.14
B25476.06EA
58.6
EUR
B25476.06
B25476.14
4-Iodcumol ≥97%
4-Iodcumol
Purity: 97%
CAS No.: 17356-09-1
Molecular Formula: C9H11I
Formula Weight: 246.09
Refractive Index: 1.5780
Boiling Point: 236-238°
MDL No.: MFCD00013711
Formel:
C₉H₁₁I Molecular Weight: 246,09 g/mol Siedepunkt: 236…238 °C Dichte: 1,525 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013711 CAS-Nummer: 17356-09-1 EINECS: 241-384-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...