2-Iod-p-toluidin
Lieferant: BIONET RESEARCH
4Y-0823.1
4Y-0823.5
4Y-0823.500
4Y-0823.10
BIOT4Y-0823.1EA
0
EUR
BIOT4Y-0823.1
BIOT4Y-0823.5
BIOT4Y-0823.500
BIOT4Y-0823.10
2-Iod-p-toluidin
2-Iod-p-toluidin
Formel:
C₇H₈IN Molecular Weight: 233.05 g/mol Siedepunkt: 279 Schmelzpunkt: 34…37 °C |
MDL:
MFCD03407391 CAS-Nummer: 29289-13-2 UN: 2811 ADR: 6.1 I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...