4-Iod-m-toluidin 98%

Lieferant: Thermo Fisher Scientific
Achtung

357800050 357800250 357801000
ACRO357800050EA 35.4 EUR
ACRO357800050 ACRO357800250 ACRO357801000
4-Iod-m-toluidin 98%
4-Iod-m-toluidin
Formel: C₇H₈IN
Molecular Weight: 233.05 g/mol
Schmelzpunkt: 50…55 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01569451
CAS-Nummer: 4949-69-3

Bestellen

Spezifikation Testergebnisse

Appearance Beige to light brown to dark grey Crystalline powder
Infrared spectrum Conforms
Melting point 50 to 55 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR