2-Methoxybenzylisothiocyanat

Lieferant: Thermo Scientific

S14972.1 S14972.10
MAYBS14972.1EA 0 EUR
MAYBS14972.1 MAYBS14972.10
2-Methoxybenzylisothiocyanat
2-Methoxybenzylisothiocyanat
Formel: C₉H₉NOS
Molecular Weight: 179.24 g/mol
MDL: MFCD00041111
CAS-Nummer: 17608-09-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR