α,α-Dimethoxytoluol 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Benzaldehyddimethylacetal
A11009.0E
A11009.22
A11009.36
A11009.0EEA
552
EUR
A11009.0E
A11009.22
A11009.36
α,α-Dimethoxytoluol 99%
α,α-Dimethoxytoluol
Purity: 99%
CAS No.: 1125-88-8
Molecular Formula: C9H12O2
Formula Weight: 152.20
Flash Point: 69°(156°F)
Refractive Index: 1.4935
Odour: Characteristic
Boiling Point: 196°
Beilstein Reference: 2044501
MDL No.: MFCD00008491
Formel:
C₉H₁₂O₂ Siedepunkt: 196 °C Dichte: 1,014 Flammpunkt: 69 °C (156 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008491 CAS-Nummer: 1125-88-8 EINECS: 214-413-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...