5-Phenyl-1-pentanol ≥97%

Lieferant: Thermo Scientific

L12070.03 L12070.06
L12070.03EA 26.4 EUR
L12070.03 L12070.06
5-Phenyl-1-pentanol ≥97%
5-Phenyl-1-pentanol

Purity: 97% CAS No.: 10521-91-2 Molecular Formula: C6H5(CH2)5OH Formula Weight: 164.25 Flash Point: 110°(230°F) Refractive Index: 1.5160 Boiling Point: 113-114°/1.5mm Beilstein Reference: 1935233 MDL No.: MFCD00002979

Formel: (C₆H₅)CH₂CH₂CH₂CH₂CH₂OH
Molecular Weight: 164,25 g/mol
Siedepunkt: 113…114 °C (1,5 mmHg)
Dichte: 0,975
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002979
CAS-Nummer: 10521-91-2
EINECS: 234-064-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR