Chloramin T (Natriumsalz) Trihydrat 98,0-103,0% ACS
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Chloramin T Natriumsalz Trihydrat, Tosylchloramidnatrium Trihydrat, N-Chlor-p-toluolsulfonamid Natriumsalz Trihydrat
Purity: 98.0-103.0%
CAS No.: 7080-50-4
Molecular Formula: CH3C6H4SO2N(Cl)Na.3H2O
Formula Weight: 281.69 (227.67anhy)
Flash Point: 192°(377°F)
Physical Form: Crystalline
Sensitivity: Moisture Sensitive
Appearance: White to pale yellow
Odour: Chlorine-like
pH: 8-10
Melting Point: 167-170°
Beilstein Reference: 3924168
Merck Reference: 14,2075
UN No.: 3263
MDL No.: MFCD00149066
Reagent for selective oxidation of methionine
Formel:
CH₃C₆H₄SO₂NClNa·3H₂O Molecular Weight: 281.69 g/mol Schmelzpunkt: 167…170 °C Dichte: 1.43 g/cm³ (20 °C) Flammpunkt: 192 °C (377 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149066 CAS-Nummer: 7080-50-4 EINECS: 204-854-7 UN: 3263 ADR: 8 III Merck Index: 14,2075 |