ω-Brom-4-phenylacetophenon ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Biphenyl-4-ylbrommethylketon, α-Brom-4-phenylacetophenon, Brommethyl-p-phenylphenylketon, 4-Phenylphenacylbromid, 2-Brom-4'-phenylacetophenon
B24298.06
B24298.14
B24298.22
B24298.06EA
26
EUR
B24298.06
B24298.14
B24298.22
ω-Brom-4-phenylacetophenon ≥97%
ω-Brom-4-phenylacetophenon
Purity: 97%
CAS No.: 135-73-9
Molecular Formula: C14H11BrO
Formula Weight: 275.15
Melting Point: 123-127°
Beilstein Reference: 513838
UN No.: 3261
MDL No.: MFCD00000202
Formel:
C₁₄H₁₁BrO Molecular Weight: 275,14 g/mol Siedepunkt: 354 °C (1013 hPa) Schmelzpunkt: 123…127 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000202 CAS-Nummer: 135-73-9 EINECS: 205-217-6 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...