4'-Phenylacetophenon ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: p-Phenylacetophenon, 1-(4-Phenylphenyl)ethanon, 4-Phenyl-1-acetylbenzol, 4-Acetylbiphenyl

A14622.14 A14622.22 A14622.36
A14622.14EA 29.8 EUR
A14622.14 A14622.22 A14622.36
4'-Phenylacetophenon ≥98%
4'-Phenylacetophenon

Purity: 98% CAS No.: 92-91-1 Molecular Formula: C14H12O Formula Weight: 196.25 Melting Point: 117-123° Boiling Point: 168°/8mm MDL No.: MFCD00008749

Formel: C₁₄H₁₂O
Molecular Weight: 196.25 g/mol
Siedepunkt: 168 °C (8 mmHg)
Schmelzpunkt: 117…123 °C
Dichte: 1.251 g/cm³ (0 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008749
CAS-Nummer: 92-91-1
EINECS: 202-202-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR