4'-Isopropylacetophenon ≥98%

Lieferant: Thermo Scientific

Synonyme: 1-(4-Isopropylphenyl)ethanon, 4-Isopropyl-1-acetylbenzol, p-Isopropylacetophenon

B20581.06 B20581.14 B20581.22
B20581.06EA 29.2 EUR
B20581.06 B20581.14 B20581.22
4'-Isopropylacetophenon ≥98%
4'-Isopropylacetophenon

Purity: 98% CAS No.: 645-13-6 Molecular Formula: C11H14O Formula Weight: 162.23 Refractive Index: 1.5230 Boiling Point: 238° MDL No.: MFCD00048297

Formel: C₁₁H₁₄O
Molecular Weight: 162,23 g/mol
Siedepunkt: 238 °C
Dichte: 0,970
Lagertemperatur: Raumtemperatur
MDL: MFCD00048297
CAS-Nummer: 645-13-6
EINECS: 211-433-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR