4-Methoxybiphenyl ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: Biphenyl-4-yl methyl ether, 4-Phenylanisole

L01260.06 L01260.14
L01260.06EA 29.6 EUR
L01260.06 L01260.14
4-Methoxybiphenyl ≥98%
4-Methoxybiphenyl

Purity: 98+% CAS No.: 613-37-6 Molecular Formula: C13H12O Formula Weight: 184.24 Melting Point: 86-90° Boiling Point: 157°/10mm MDL No.: MFCD00014897

Formel: C₁₃H₁₂O
Molecular Weight: 184.24 g/mol
Siedepunkt: 157 °C (10 mmHg)
Schmelzpunkt: 86…90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00014897
CAS-Nummer: 613-37-6
EINECS: 210-339-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR