4-Methoxybiphenyl

Lieferant: CHEMPUR

Synonyme: Biphenyl-4-yl methyl ether, 4-Phenylanisole

SEW03888.250 SEW03888.1
CHMPSEW03888.250EA 0 EUR
CHMPSEW03888.250 MAYBSEW03888.1
4-Methoxybiphenyl
4-Methoxybiphenyl
Formel: C₁₃H₁₂O
Molecular Weight: 184,24 g/mol
MDL: MFCD00014897
CAS-Nummer: 613-37-6
EINECS: 210-339-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR