Tetrafluor-p-benzochinon

Lieferant: FLUOROCHEM

Synonyme: Tetrafluoroquinone, 2,3,5,6-Tetrafluoro-1,4-benzoquinone, Tetrafluoro-1,4-benzoquinone, p-Fluoranil, 4-Fluoranil

003313-1G 003313-5G 003313-25G
FLUO003313-1GEA 63.9 EUR
FLUO003313-1G FLUO003313-5G FLUO003313-25G
Tetrafluor-p-benzochinon
Tetrafluor-p-benzochinon
Formel: C₆F₄O₂
Molecular Weight: 180.06 g/mol
Siedepunkt: 133
MDL: MFCD00001592
CAS-Nummer: 527-21-9
EINECS: 208-411-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR