Homopiperazin ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 1,4-Diazacycloheptan, Perhydro-1,4-diazepin

A15751.14 A15751.22
A15751.14EA 54 EUR
A15751.14 A15751.22
Homopiperazin ≥98%
Homopiperazin

Purity: 98% CAS No.: 505-66-8 Molecular Formula: C5H12N2 Formula Weight: 100.17 Flash Point: 64°(147°F) Odour: Amine-like Boiling Point: 168-170° Beilstein Reference: 102711 UN No.: 3259 MDL No.: MFCD00006933

Formel: C₅H₁₂N₂
Molecular Weight: 100,16 g/mol
Siedepunkt: 168…170 °C
Schmelzpunkt: 38…44 °C
Dichte: 0,95
Flammpunkt: 64 °C (147 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006933
CAS-Nummer: 505-66-8
EINECS: 208-016-1
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR