1-(2-Methoxyethyl)piperazin
Lieferant: FLUOROCHEM
008907-1G
008907-5G
FLUO008907-1GEA
31.1
EUR
FLUO008907-1G
FLUO008907-5G
1-(2-Methoxyethyl)piperazin
1-(2-Methoxyethyl)piperazin
Formel:
C₇H₁₆N₂O Molecular Weight: 144.22 g/mol |
MDL:
MFCD00191214 CAS-Nummer: 13484-40-7 UN: 3267 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...