1-(4-Bromphenyl)piperazin

Lieferant: FLUOROCHEM

019213-1G 019213-5G
FLUO019213-1GEA 0 EUR
FLUO019213-1G FLUO019213-5G
1-(4-Bromphenyl)piperazin
1-(4-Bromphenyl)piperazin
Formel: C₁₀H₁₃BrN₂
Molecular Weight: 241,13 g/mol
MDL: MFCD00130198
CAS-Nummer: 66698-28-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR