1-(3,5-Dichlorphenyl)piperazin

Lieferant: FLUOROCHEM

009769-1G 009769-5G
FLUO009769-1GEA 0 EUR
FLUO009769-1G FLUO009769-5G
1-(3,5-Dichlorphenyl)piperazin
1-(3,5-Dichlorphenyl)piperazin
Formel: C₁₀H₁₂Cl₂N₂
Molecular Weight: 231.12 g/mol
MDL: MFCD00040763
CAS-Nummer: 55827-50-4
EINECS: 259-841-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR