1-(4-Nitrophenyl)piperazin 98%
Lieferant: Thermo Scientific
|
Gefahr
|
L18700.14
L18700.06
L18700.22
L18700.14EA
90.2
EUR
L18700.14
L18700.06
L18700.22
1-(4-Nitrophenyl)piperazin 98%
1-(4-Nitrophenyl)piperazin
Purity: 98+%
CAS No.: 6269-89-2
Molecular Formula: C10H13N3O2
Formula Weight: 207.23
Melting Point: 129-133°
UN No.: 2811
MDL No.: MFCD00005961
Formel:
C₁₀H₁₃N₃O₂ Molecular Weight: 207,23 g/mol Schmelzpunkt: 129…133 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005961 CAS-Nummer: 6269-89-2 EINECS: 228-443-7 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...