1-(3-Methoxyphenyl)piperazin

Lieferant: FLUOROCHEM

009802-5G 009802-25G
FLUO009802-5GEA 0 EUR
FLUO009802-5G FLUO009802-25G
1-(3-Methoxyphenyl)piperazin
1-(3-Methoxyphenyl)piperazin
Formel: C₁₁H₁₆N₂O
Molecular Weight: 192,26 g/mol
Siedepunkt: 130 °C (0,5 mmHg)
Dichte: 1,082 g/cm³ (20 °C)
MDL: MFCD00040733
CAS-Nummer: 16015-71-7
EINECS: 240-154-8
UN: 3267
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR