1-(4-Methoxyphenyl)piperazin

Lieferant: FLUOROCHEM

009803-5G 009803-25G
FLUO009803-5GEA 0 EUR
FLUO009803-5G FLUO009803-25G
1-(4-Methoxyphenyl)piperazin
1-(4-Methoxyphenyl)piperazin
Formel: C₁₁H₁₆N₂O
Molecular Weight: 192,26 g/mol
Siedepunkt: 118 °C (0,3 mmHg)
Schmelzpunkt: 261…262 °C
MDL: MFCD00040736
CAS-Nummer: 38212-30-5
EINECS: 253-829-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR