1-(2-Hydroxyphenyl)piperazin ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
o-(1-Piperazinyl)phenol
B20252.09
B20252.18
B20252.09EA
39.5
EUR
B20252.09
B20252.18
1-(2-Hydroxyphenyl)piperazin ≥98%
1-(2-Hydroxyphenyl)piperazin
Purity: 98%
CAS No.: 1011-17-2
Molecular Formula: C10H14N2O
Formula Weight: 178.24
Melting Point: 122-127°
Beilstein Reference: 10646
UN No.: 3263
MDL No.: MFCD00190246
Formel:
C₁₀H₁₄N₂O Molecular Weight: 178,23 g/mol Siedepunkt: 318 °C (1013 hPa) Schmelzpunkt: 122…127 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00190246 CAS-Nummer: 1011-17-2 EINECS: 213-782-5 UN: 3263 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...