1-(3-Hydroxyphenyl)piperazin ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
m-(1-Piperazinyl)phenol
L18702.06
L18702.14
L18702.06EA
27.8
EUR
L18702.06
L18702.14
1-(3-Hydroxyphenyl)piperazin ≥97%
1-(3-Hydroxyphenyl)piperazin
Purity: 98%
CAS No.: 59817-32-2
Molecular Formula: C10H14N2O
Formula Weight: 178.24
Melting Point: 205-208°
Beilstein Reference: 4248583
UN No.: 3263
MDL No.: MFCD00052896
Formel:
C₁₀H₁₄N₂O Molecular Weight: 178,23 g/mol Schmelzpunkt: 205…208 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052896 CAS-Nummer: 59817-32-2 UN: 3263 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...