1-(3-Hydroxyphenyl)piperazin
Lieferant: FLUOROCHEM
Synonyme:
m-(1-Piperazinyl)phenol
009796-10G
009796-50G
009796-1G
009796-5G
FLUO009796-10GEA
0
EUR
FLUO009796-10G
FLUO009796-50G
FLUO009796-1G
FLUO009796-5G
1-(3-Hydroxyphenyl)piperazin
1-(3-Hydroxyphenyl)piperazin
Formel:
C₁₀H₁₄N₂O Schmelzpunkt: 210…212 °C |
MDL:
MFCD00052896 CAS-Nummer: 59817-32-2 UN: 3263 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...