1-(4-Hydroxyphenyl)piperazin ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
p-(1-Piperazinyl)phenol
B20122.18
B20122.30
B20122.09
B20122.18EA
145
EUR
B20122.18
B20122.30
B20122.09
1-(4-Hydroxyphenyl)piperazin ≥98%
1-(4-Hydroxyphenyl)piperazin
Purity: 98%
CAS No.: 56621-48-8
Molecular Formula: C10H14N2O
Formula Weight: 178.24
Melting Point: 218-224°
Beilstein Reference: 150964
UN No.: 3263
MDL No.: MFCD00066156
Formel:
C₁₀H₁₄N₂O Molecular Weight: 178,23 g/mol Schmelzpunkt: 218…224 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066156 CAS-Nummer: 56621-48-8 EINECS: 260-289-6 UN: 3263 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...