4-(Dimethylamino)antipyrin 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2,3-Dimethyl-4-(dimethylamino)-1-phenyl-1,2-diazacyclopent-3-en-5-on, 4-(Dimethylamino)phenazon

115740050 115741000 115740010 115740250
ACRO115740050EA 21.6 EUR
ACRO115740050 ACRO115741000 ACRO115740010 ACRO115740250
4-(Dimethylamino)antipyrin 97%
4-(Dimethylamino)antipyrin
Formel: C₁₃H₁₇N₃O
Molecular Weight: 231.3 g/mol
Schmelzpunkt: 105…110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003142
CAS-Nummer: 58-15-1
EINECS: 200-365-8
UN: 2811
ADR: 6.1 III
Merck Index: 12,00495

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder or crystals
Infrared spectrum Conforms
Melting point 105 to 110 °C
Titration with HClO4 ≥96.0 %
Specific optical rotation -15° to -18° (20 °C, 589 nm) (c=10, EtOH)

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR