4-(Dimethylamino)antipyrin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2,3-Dimethyl-4-(dimethylamino)-1-phenyl-1,2-diazacyclopent-3-en-5-on, 4-(Dimethylamino)phenazon
M-Clarity™ Qualitätsstufe = MQ200
D8015-100G
D8015-25G
SIALD8015-100GEA
215
EUR
SIALD8015-100G
SIALD8015-25G
4-(Dimethylamino)antipyrin, Sigma-Aldrich®
4-(Dimethylamino)antipyrin
Formel:
C₁₃H₁₇N₃O Molecular Weight: 231,3 g/mol Schmelzpunkt: 134…135 °C |
MDL:
MFCD00003142 CAS-Nummer: 58-15-1 EINECS: 200-365-8 UN: 2811 ADR: 6.1,III Merck Index: 12,00495 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...