4-Aminoantipyrin 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-on, 4-Aminophenazon

103150250 103151000 103155000 103150025
ACRO103150250EA 34.7 EUR
ACRO103150250 ACRO103151000 ACRO103155000 ACRO103150025
4-Aminoantipyrin 98%
4-Aminoantipyrin
Formel: C₁₁H₁₃N₃O
Molecular Weight: 203,24 g/mol
Schmelzpunkt: 105,5…110 °C
Dichte: 0,8 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00003145
CAS-Nummer: 83-07-8
EINECS: 201-452-3
Merck Index: 13,00596

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to yellow-brown Powder or crystals
Infrared spectrum Conforms
Melting point 105 to 110 °C
Loss on drying ≤1.5 %
Sulfated ash ≤0.1 %
Solubility (5 % in water) Clear
Titration with HClO4 ≥97.5 % (On dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR