4-Aminoantipyrin 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-on, 4-Aminophenazon

A13846.0E A13846.22 A13846.36
A13846.0EEA 1069 EUR
A13846.0E A13846.22 A13846.36
4-Aminoantipyrin 97%
4-Aminoantipyrin

Purity: 97% CAS No.: 83-07-8 Molecular Formula: C11H13N3O Formula Weight: 203.25 Physical Form: Powder Appearance: Yellow to pale brown Melting Point: 106-110° Beilstein Reference: 181635 Merck Reference: 14,591 MDL No.: MFCD00003145

Formel: C₁₁H₁₃N₃O
Molecular Weight: 203,24 g/mol
Schmelzpunkt: 106…110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003145
CAS-Nummer: 83-07-8
EINECS: 201-452-3
Merck Index: 14,591

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR