Phenothiazin 99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
10H-Phenothiazine
130210010
130215000
130210050
ACRO130210010EA
95.5
EUR
ACRO130210010
ACRO130215000
ACRO130210050
Phenothiazin 99%
Phenothiazin
Formel:
C₁₂H₉NS Molecular Weight: 199,28 g/mol Siedepunkt: 371 °C Schmelzpunkt: 185 °C Dichte: 1,34 g/cm³ (20 °C) Flammpunkt: >100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005015 CAS-Nummer: 92-84-2 EINECS: 202-196-5 UN: 3077 ADR: 9,III Merck Index: 12,07404 |
Spezifikation Testergebnisse
Appearance | Yellow Beads or prills |
Infrared spectrum | Conforms |
Melting point | ≥184 °C |
HPLC | ≥98.5 % |
UV | at 260 nm A: ≤0.02 |
Titration after Ion exchange | ≥99.0 % (on dry substance) |
Membrane filtration test | (on 0.2 M soln.) Passes test |
Suitability | (HPLC gradient) Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...